ENA428073271 Enamine

3-chloro-5H,6H,7H-cyclopenta[c]pyridine-4-carbonitrile

  • MDL number MFCD19690416
  • Cas Number : 103589-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-106
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-178115

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178115
Availability details for
Quantity
  • Availability
  • Add to Cart