ENA428073273 Enamine

ethyl 5-amino-1-(pyridin-4-yl)-1H-imidazole-4-carboxylate

  • MDL number MFCD16529586
  • Cas Number : 1280582-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178117

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178117
Availability details for
Quantity
  • Availability
  • Add to Cart