ENA428073274 Enamine

2-chloro-1-(1,3-thiazol-4-yl)ethan-1-one

  • MDL number MFCD19232564
  • Cas Number : 52540-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178118

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178118
Availability details for
Quantity
  • Availability
  • Add to Cart