ENA428073284 Enamine

7-bromo-1-benzothiophene-3-carboxylic acid

  • MDL number MFCD18451782
  • Cas Number : 19075-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178136

Safety Information

msds https://www.enaminestore.com/catalog/EN300-178136
Availability details for
Quantity
  • Availability
  • Add to Cart