ENA428073287 Enamine

2-{[(5-bromothiophen-2-yl)methyl]amino}ethan-1-ol

  • MDL number MFCD11175358
  • Cas Number : 856371-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178153

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178153
Availability details for
Quantity
  • Availability
  • Add to Cart