ENA428073292 Enamine

(4-cyclohexyl-1,3-thiazol-2-yl)methanol

  • MDL number MFCD17251226
  • Cas Number : 1342526-88-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178175

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178175
Availability details for
Quantity
  • Availability
  • Add to Cart