ENA428073295 Enamine

1-{2-methyl-[1,2,4]triazolo[1,5-a]pyrazin-5-yl}piperazine

  • MDL number MFCD28023724
  • Cas Number : 1697650-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-159353

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-159353
Availability details for
Quantity
  • Availability
  • Add to Cart