ENA428073298 Enamine

N,N-dimethyl-4-(methylamino)benzene-1-sulfonamide

  • MDL number MFCD11207401
  • Cas Number : 77812-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-178208

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178208
Availability details for
Quantity
  • Availability
  • Add to Cart