ENA428073305 Enamine

1-[5-(butan-2-yl)-4H-1,2,4-triazol-3-yl]ethan-1-amine

  • MDL number MFCD18872137
  • Cas Number : 1343630-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178226

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178226
Availability details for
Quantity
  • Availability
  • Add to Cart