ENA428073307 Enamine

6-(3-ethoxyphenyl)pyridin-2-amine

  • MDL number MFCD18510662
  • Cas Number : 1125416-16-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178237

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178237
Availability details for
Quantity
  • Availability
  • Add to Cart