ENA428073310 Enamine

3,7-bis(trifluoromethyl)-1,2-dihydroquinoxalin-2-one

  • MDL number MFCD28023661
  • Cas Number : 231607-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-178243

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178243
Availability details for
Quantity
  • Availability
  • Add to Cart