ENA428073318 Enamine

2-chloro-2-fluoro-2-phenylacetic acid

  • MDL number MFCD20644756
  • Cas Number : 134918-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-178281

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178281
Availability details for
Quantity
  • Availability
  • Add to Cart