ENA428073320 Enamine

3-(butan-2-yl)cyclopentan-1-one

  • MDL number MFCD21929742
  • Cas Number : 91523-76-1
  • Molecular Weight : 140.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-178285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178285
Availability details for
Quantity
  • Availability
  • Add to Cart