ENA428073333 Enamine

2,3-dihydro-1,4-benzoxathiine-2-carbonitrile

  • MDL number MFCD28064179
  • Cas Number : 51263-42-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178321

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178321
Availability details for
Quantity
  • Availability
  • Add to Cart