ENA428073338 Enamine

3-[(4-chlorophenyl)methyl]piperidin-2-one

  • MDL number MFCD11848670
  • Cas Number : 1260763-19-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178373

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178373
Availability details for
Quantity
  • Availability
  • Add to Cart