ENA428073344 Enamine

3-(2-methylpropyl)cyclopentan-1-one

  • MDL number MFCD20361790
  • Cas Number : 53002-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178385

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178385
Availability details for
Quantity
  • Availability
  • Add to Cart