ENA428073547 Enamine

5-methyl-6-(1H-pyrazol-1-yl)pyridine-3-carbonitrile

  • MDL number MFCD28064181
  • Cas Number : 1803570-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178418

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178418
Availability details for
Quantity
  • Availability
  • Add to Cart