ENA428073555 Enamine

butane-2,3-diamine dihydrochloride

  • MDL number MFCD00035525
  • Cas Number : 55536-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 299-303
Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-178572

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178572
Availability details for
Quantity
  • Availability
  • Add to Cart