ENA428073559 Enamine

3-(2,3-dihydro-1H-isoindol-2-yl)-2-hydroxy-2-methylpropanoic acid hydrochloride

  • MDL number MFCD28064183
  • Cas Number : 1803592-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178613

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178613
Availability details for
Quantity
  • Availability
  • Add to Cart