ENA428073569 Enamine

[2,5-difluoro-4-(trifluoromethyl)phenyl]methanol

  • MDL number MFCD28063904
  • Cas Number : 1803598-53-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-178864

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178864
Availability details for
Quantity
  • Availability
  • Add to Cart