ENA428073570 Enamine

1,3-dibromo-2-(difluoromethoxy)-4-nitrobenzene

  • MDL number MFCD28063907
  • Cas Number : 1803570-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178869

Safety Information

msds https://www.enaminestore.com/catalog/EN300-178869
Availability details for
Quantity
  • Availability
  • Add to Cart