ENA428073571 Enamine

7-(aminomethyl)-6-oxaspiro[3.4]octan-5-one hydrochloride

  • MDL number MFCD28063909
  • Cas Number : 1803602-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178879

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178879
Availability details for
Quantity
  • Availability
  • Add to Cart