ENA428073572 Enamine

4-(2-bromoethyl)-1,4-oxazepane hydrochloride

  • MDL number MFCD28063910
  • Cas Number : 1803585-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178900

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178900
Availability details for
Quantity
  • Availability
  • Add to Cart