ENA428073573 Enamine

3-(4-bromophenyl)-5-(pyrrolidin-2-yl)-1,2,4-oxadiazole hydrochloride

  • MDL number MFCD28063911
  • Cas Number : 1803608-62-9
  • Molecular Weight : 330.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-178901

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178901
Availability details for
Quantity
  • Availability
  • Add to Cart