ENA428073577 Enamine

6,6,6-trifluorohexan-2-amine hydrochloride

  • MDL number MFCD28063917
  • Cas Number : 1803595-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178980

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178980
Availability details for
Quantity
  • Availability
  • Add to Cart