ENA428073579 Enamine

2-(2,5-dimethylpiperidin-1-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD28063922
  • Cas Number : 1803605-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-179041

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179041
Availability details for
Quantity
  • Availability
  • Add to Cart