ENA428073582 Enamine

[(5-bromothiophen-2-yl)methyl](2-methoxyethyl)amine hydrochloride

  • MDL number MFCD28063925
  • Cas Number : 1645414-72-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179049

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179049
Availability details for
Quantity
  • Availability
  • Add to Cart