ENA428073587 Enamine

sodium 2-cyano-4-phenylbutanoate

  • MDL number MFCD28063931
  • Cas Number : 1798750-53-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-179077

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179077
Availability details for
Quantity
  • Availability
  • Add to Cart