ENA428073591 Enamine

1,4-dimethyl 2-(difluoromethoxy)butanedioate

  • MDL number MFCD28063935
  • Cas Number : 1803598-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-179084
Availability details for
Quantity
  • Availability
  • Add to Cart