ENA428073593 Enamine

2,4-dimethyl-3,5-dinitrobenzene-1-sulfonyl fluoride

  • MDL number MFCD28063938
  • Cas Number : 1803592-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179176

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179176
Availability details for
Quantity
  • Availability
  • Add to Cart