ENA428073597 Enamine

5-(piperidin-4-yl)isoquinoline dihydrochloride

  • MDL number MFCD28063943
  • Cas Number : 1803587-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-179211

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179211
Availability details for
Quantity
  • Availability
  • Add to Cart