ENA428073599 Enamine

[1-(aminomethyl)-2-(4-fluorophenyl)cyclopropyl]methanol hydrochloride

  • MDL number MFCD28063946
  • Cas Number : 1798730-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-179251

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179251
Availability details for
Quantity
  • Availability
  • Add to Cart