ENA428073602 Enamine

1-(3-chloro-4-methanesulfonylphenyl)methanamine hydrochloride

  • MDL number MFCD28063724
  • Cas Number : 106737-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179306

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179306
Availability details for
Quantity
  • Availability
  • Add to Cart