ENA428073603 Enamine

2-bromo-5-cyclopropyl-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD28063725
  • Cas Number : 1694180-93-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179341

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179341
Availability details for
Quantity
  • Availability
  • Add to Cart