ENA428073606 Enamine

6-(1,4-diazepan-1-yl)-9H-purine trihydrochloride

  • MDL number MFCD28063731
  • Cas Number : 1803596-68-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 3HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179378

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179378
Availability details for
Quantity
  • Availability
  • Add to Cart