ENA428073608 Enamine

(1R)-1-cyclobutylethan-1-amine hydrochloride

  • MDL number MFCD28063736
  • Cas Number : 677743-79-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-179429

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179429
Availability details for
Quantity
  • Availability
  • Add to Cart