ENA428073609 Enamine

2-[3-(methoxymethyl)piperidin-1-yl]ethan-1-amine dihydrochloride

  • MDL number MFCD28063739
  • Cas Number : 1645384-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179436

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179436
Availability details for
Quantity
  • Availability
  • Add to Cart