ENA428073615 Enamine

ethyl imidazo[1,2-c]pyrimidine-7-carboxylate

  • MDL number MFCD28063748
  • Cas Number : 588720-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-179519

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179519
Availability details for
Quantity
  • Availability
  • Add to Cart