ENA428073616 Enamine

sodium 1-{1-[(tert-butoxy)carbonyl]piperidin-3-yl}-1H-1,2,4-triazole-3-carboxylate

  • MDL number MFCD28063750
  • Cas Number : 1803590-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179570

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179570
Availability details for
Quantity
  • Availability
  • Add to Cart