ENA428073622 Enamine

2,2,3-trimethylbutane-1-sulfonyl chloride

  • MDL number MFCD28063757
  • Cas Number : 1823344-23-5
  • Molecular Weight : 198.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,2,3-trimethylbutane-1-sulfonyl chloride
Inchi Code 1S/C7H15ClO2S/c1-6(2)7(3,4)5-11(8,9)10/h6H,5H2,1-4H3
Inchi Key BUYBIZNSGFHMJB-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-179653

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179653
Availability details for
Quantity
  • Availability
  • Add to Cart