ENA428073623 Enamine

2,2,3-trimethylbutane-1-sulfonamide

  • MDL number MFCD28063758
  • Cas Number : 1823341-54-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179654

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179654
Availability details for
Quantity
  • Availability
  • Add to Cart