ENA428073624 Enamine

4-{[(tert-butyldimethylsilyl)oxy]methyl}cyclohexan-1-ol

  • MDL number MFCD28063759
  • Cas Number : 117107-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-179664

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179664
Availability details for
Quantity
  • Availability
  • Add to Cart