ENA428073629 Enamine

1-(2-oxopropyl)-1,2-dihydropyridin-2-one hydrochloride

  • MDL number MFCD28063768
  • Cas Number : 1788611-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179970

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179970
Availability details for
Quantity
  • Availability
  • Add to Cart