ENA428073630 Enamine

2-benzyl-5H,6H,7H,8H-imidazo[1,2-a]pyrazine dihydrochloride

  • MDL number MFCD28063769
  • Cas Number : 1803605-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179978

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179978
Availability details for
Quantity
  • Availability
  • Add to Cart