ENA428073641 Enamine

3-amino-6-(dimethylamino)-1-methyl-2,3-dihydro-1H-indol-2-one dihydrochloride

  • MDL number MFCD28063963
  • Cas Number : 1786215-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180471

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180471
Availability details for
Quantity
  • Availability
  • Add to Cart