ENA428073643 Enamine

7-(morpholine-4-sulfonyl)-1,2,3,4-tetrahydroisoquinolin-1-one

  • MDL number MFCD28063966
  • Cas Number : 1803598-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180503

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180503
Availability details for
Quantity
  • Availability
  • Add to Cart