ENA428073654 Enamine

4-amino-3-(propan-2-yloxy)phenol hydrochloride

  • MDL number MFCD28063985
  • Cas Number : 1803606-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180741

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180741
Availability details for
Quantity
  • Availability
  • Add to Cart