ENA428073661 Enamine

2,2,2-trichloro-1-{1-[(thiophen-2-yl)methyl]-1H-pyrrol-2-yl}ethan-1-one

  • MDL number MFCD28064031
  • Cas Number : 1803589-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180898

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180898
Availability details for
Quantity
  • Availability
  • Add to Cart