ENA428073664 Enamine

3-(3-methyl-1,2,4-oxadiazol-5-yl)pyrrolidin-3-ol hydrochloride

  • MDL number MFCD28064034
  • Cas Number : 1803606-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180906

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180906
Availability details for
Quantity
  • Availability
  • Add to Cart