ENA428073666 Enamine

ethyl 3-(piperidin-4-yl)-1H-1,2,4-triazole-5-carboxylate hydrochloride

  • MDL number MFCD28064036
  • Cas Number : 1795504-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180940

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180940
Availability details for
Quantity
  • Availability
  • Add to Cart